Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014901.1 | NP_001014901.1 | 2 - 3 | 1887 | 87 | |
GenBank mRNA | BT020733.1 | - | 2 - 3 | 1887 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Bos taurus Probe spans exons |