Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015596.1 | NP_001015596.1 | 3 - 4 | 461 | 68 | |
GenBank mRNA | BC112869.1 | - | 3 - 4 | 469 | 68 | |
BT020676.1 | - | 3 - 4 | 461 | 68 | ||
BT026291.1 | - | 3 - 4 | 456 | 68 | ||
BT029889.1 | - | 3 - 4 | 456 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |