Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015613.1 | NP_001015613.1 | 4 - 5 | 524 | 82 | |
GenBank mRNA | BC102333.1 | - | 4 - 5 | 563 | 82 | |
BT020708.1 | - | 4 - 5 | 524 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |