Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPUSD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015616.1 | NP_001015616.1 | 2 - 3 | 358 | 129 | |
GenBank mRNA | BC134590.1 | - | 2 - 3 | 380 | 129 | |
BT020717.1 | - | 2 - 3 | 358 | 129 | ||
BT020773.1 | - | 2 - 3 | 359 | 129 | ||
BT020778.1 | - | 2 - 3 | 358 | 129 | ||
BT021642.1 | - | 2 - 3 | 358 | 129 | ||
BT021761.1 | - | 2 - 3 | 358 | 129 | ||
BT025340.1 | - | 2 - 3 | 344 | 129 | ||
BT025483.1 | - | 2 - 3 | 350 | 129 | ||
BT026244.1 | - | 2 - 3 | 369 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |