Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIAPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015659.1 | NP_001015659.1 | 3 - 4 | 371 | 84 | |
GenBank mRNA | BT020642.1 | - | 3 - 4 | 371 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |