Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017933.1 | NP_001017933.1 | 1 - 2 | 199 | 72 | |
GenBank mRNA | BC122698.1 | - | 1 - 2 | 142 | 72 | |
BT020635.1 | - | 1 - 2 | 199 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |