Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025207.1 | NP_001020378.1 | 1 - 2 | 417 | 101 | |
GenBank mRNA | BC134761.1 | - | 1 - 2 | 483 | 101 | |
BT020722.1 | - | 1 - 2 | 417 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |