Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031760.1 | NP_001026930.1 | 7 - 8 | 751 | 135 | |
GenBank mRNA | BT020672.1 | - | 7 - 8 | 751 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |