Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034034.2 | NP_001029206.1 | 1 - 2 | 50 | 67 | |
GenBank mRNA | AB098979.1 | - | 1 - 2 | 37 | 67 | |
AB098985.1 | - | 1 - 2 | 37 | 67 | ||
BC102589.1 | - | 1 - 2 | 51 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |