Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034295.2 | NP_001029467.1 | 5 - 6 | 590 | 67 | |
GenBank mRNA | BC103176.1 | - | 5 - 6 | 591 | 67 | |
GQ166651.1 | - | 3 - 4 | 504 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |