Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034309.2 | NP_001029481.1 | 4 - 5 | 790 | 143 | |
GenBank mRNA | BC103199.1 | - | 4 - 5 | 791 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |