Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034340.2 | NP_001029512.1 | 2 - 3 | 497 | 115 | |
GenBank mRNA | BC102934.1 | - | 2 - 3 | 498 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |