Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034368.1 | NP_001029540.1 | 6 - 7 | 975 | 69 | |
GenBank mRNA | BC114030.1 | - | 6 - 7 | 948 | 69 | |
BT020710.1 | - | 6 - 7 | 975 | 69 | ||
BT029880.1 | - | 6 - 7 | 976 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |