Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034458.1 | NP_001029630.1 | 1 | 241 | 122 | |
GenBank mRNA | BC103193.1 | - | 1 | 241 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |