Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034661.2 | NP_001029833.1 | 5 - 6 | 870 | 99 | |
GenBank mRNA | BC102522.1 | - | 5 - 6 | 872 | 99 | |
DQ319074.1 | - | 4 - 5 | 682 | 99 | ||
DQ407276.1 | - | 4 - 5 | 682 | 99 | ||
DQ471669.1 | - | 4 - 5 | 689 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |