Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034750.2 | NP_001029922.1 | 3 - 4 | 389 | 106 | |
GenBank mRNA | BC103191.1 | - | 3 - 4 | 390 | 106 | |
BC142297.1 | - | 6 - 7 | 965 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |