Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035050.2 | NP_001030222.1 | 2 - 3 | 279 | 89 | |
GenBank mRNA | BC103132.1 | - | 2 - 3 | 281 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |