Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035334.1 | NP_001030411.1 | 2 - 3 | 381 | 98 | |
GenBank mRNA | BC102233.1 | - | 2 - 3 | 381 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |