Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLVAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035353.2 | NP_001030430.1 | 4 - 5 | 1292 | 96 | |
GenBank mRNA | BC102834.1 | - | 4 - 5 | 1295 | 96 | |
BT025486.1 | - | 4 - 5 | 1243 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |