Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038022.1 | NP_001033111.1 | 7 - 8 | 568 | 77 | |
GenBank mRNA | BC109716.1 | - | 7 - 8 | 573 | 77 | |
BC153837.1 | - | 6 - 7 | 975 | 77 | ||
X65020.1 | - | 568 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |