Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C21H15orf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075386.2 | NP_001068854.1 | 3 - 4 | 341 | 99 | |
GenBank mRNA | BC103179.1 | - | 3 - 4 | 343 | 99 | |
BC149508.1 | - | 3 - 4 | 1010 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |