Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075735.1 | NP_001069203.1 | 89 | 72 | ||
GenBank mRNA | BC103254.1 | - | 89 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus |