Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075735.1 | NP_001069203.1 | 159 | 124 | ||
GenBank mRNA | BC103254.1 | - | 159 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus |