Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173897.1 | NP_776322.1 | 4 - 5 | 454 | 78 | |
GenBank mRNA | BC142534.1 | - | 4 - 5 | 469 | 78 | |
U58870.1 | - | 4 - 5 | 454 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |