Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173903.4 | NP_776328.2 | 7 - 8 | 1455 | 66 | |
GenBank mRNA | BC142181.1 | - | 7 - 8 | 1321 | 66 | |
CB437028.1 | - | 3 - 4 | 414 | 66 | ||
EW682509.1 | - | 2 - 3 | 182 | 66 | ||
X04411.1 | - | 1086 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |