Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173921.2 | NP_776346.1 | 3 - 4 | 375 | 66 | |
GenBank mRNA | EU276069.1 | - | 3 - 4 | 345 | 66 | |
M77120.1 | - | 3 - 4 | 375 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Bos taurus Probe spans exons |