Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173938.3 | NP_776363.2 | 4 - 5 | 296 | 78 | |
GenBank mRNA | AF068196.1 | - | 3 - 4 | 296 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |