Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173954.2 | NP_776379.2 | 1 - 2 | 116 | 140 | |
GenBank mRNA | J00024.1 | - | 1 - 2 | 81 | 140 | |
M25082.1 | - | 1 - 2 | 116 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |