Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173974.3 | NP_776399.2 | 5 - 6 | 849 | 65 | |
GenBank mRNA | BC146035.1 | - | 5 - 6 | 851 | 65 | |
BT021613.1 | - | 5 - 6 | 773 | 65 | ||
U05775.1 | - | 5 - 6 | 966 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() |