Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174008.1 | NP_776433.1 | 1 - 2 | 89 | 127 | |
GenBank mRNA | AF141313.1 | - | 1 - 2 | 89 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Bos taurus Inventoried Probe spans exons |