Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174048.2 | NP_776473.1 | 6 - 7 | 640 | 74 | |
GenBank mRNA | BC149525.1 | - | 6 - 7 | 657 | 74 | |
M80838.1 | - | 6 - 7 | 645 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |