Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174067.2 | NP_776492.1 | 3 - 4 | 331 | 86 | |
GenBank mRNA | AF350329.1 | - | 3 - 4 | 331 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |