Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174103.2 | NP_776528.1 | 7 - 8 | 885 | 84 | |
GenBank mRNA | L26319.1 | - | 7 - 8 | 885 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |