Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPFF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174123.3 | NP_776548.1 | 2 - 3 | 239 | 54 | |
GenBank mRNA | AF148699.1 | - | 2 - 3 | 240 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |