Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174239.2 | NP_776664.1 | 10 - 11 | 1236 | 123 | |
GenBank mRNA | BC105193.1 | - | 10 - 11 | 123 | ||
BT030667.1 | - | 10 - 11 | 1236 | 123 | ||
L36128.1 | - | 10 - 11 | 1236 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |