Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174254.2 | NP_776679.1 | 4 - 5 | 448 | 94 | |
GenBank mRNA | AF169150.1 | - | 3 - 4 | 271 | 94 | |
AF169151.1 | - | 4 - 5 | 448 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |