Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174272.2 | NP_776697.1 | 6 - 7 | 625 | 83 | |
GenBank mRNA | X02597.1 | - | 625 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |