Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174272.2 | NP_776697.1 | 8 - 9 | 931 | 72 | |
GenBank mRNA | X02597.1 | - | 931 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |