Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174312.2 | NP_776737.1 | 3 - 4 | 574 | 79 | |
GenBank mRNA | AF030029.1 | - | 3 - 4 | 574 | 79 | |
AF030030.1 | - | 2 - 3 | 433 | 79 | ||
BC151475.1 | - | 3 - 4 | 549 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |