Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174321.2 | NP_776746.1 | 2 - 3 | 409 | 72 | |
GenBank mRNA | AF268375.1 | - | 2 - 3 | 409 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |