Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174323.2 | NP_776748.1 | 5 - 6 | 1304 | 104 | |
GenBank mRNA | D90335.1 | - | 5 - 6 | 1304 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |