Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174326.2 | NP_776751.1 | 3 - 4 | 476 | 109 | |
GenBank mRNA | M11116.1 | - | 3 - 4 | 476 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |