Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174355.2 | NP_776780.1 | 4 - 5 | 324 | 92 | |
GenBank mRNA | EU276075.1 | - | 4 - 5 | 324 | 92 | |
U14416.1 | - | 4 - 5 | 324 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |