Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174373.2 | NP_776798.1 | 1 - 2 | 182 | 75 | |
GenBank mRNA | U95369.1 | - | 1 - 2 | 182 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |