Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCOCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174400.2 | NP_776825.1 | 6 - 7 | 701 | 118 | |
GenBank mRNA | AB008852.1 | - | 6 - 7 | 701 | 118 | |
BC146237.1 | - | 6 - 7 | 723 | 118 | ||
BC151569.1 | - | 6 - 7 | 723 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |