Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174403.2 | NP_776828.1 | 6 - 7 | 1261 | 80 | |
GenBank mRNA | D13569.1 | - | 6 - 7 | 1261 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |