Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAUR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174423.3 | NP_776848.1 | 4 - 5 | 488 | 69 | |
GenBank mRNA | AF144762.1 | - | 3 - 4 | 391 | 69 | |
BC134754.1 | - | 4 - 5 | 495 | 69 | ||
BC142002.1 | - | 4 - 5 | 497 | 69 | ||
L03545.1 | - | 4 - 5 | 483 | 69 | ||
S70635.1 | - | 4 - 5 | 488 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |