Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174433.2 | NP_776858.1 | 4 - 5 | 694 | 137 | |
GenBank mRNA | AF305563.1 | - | 4 - 5 | 694 | 137 | |
BC109824.1 | - | 4 - 5 | 661 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |