Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174434.3 | NP_776859.1 | 1 - 2 | 981 | 58 | |
GenBank mRNA | AF163568.1 | - | 1 - 2 | 983 | 58 | |
BC149945.1 | - | 2 - 3 | 1144 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |