Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174455.3 | NP_776880.1 | 1 - 2 | 49 | 60 | |
GenBank mRNA | BC102108.1 | - | 1 - 2 | 48 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |